Norwell Dipstik D8653274 PTFE Drum And Tank Sampler, 7' Long
Representative image only.
Norwell Dipstik Mfr # D8653274 – Item # EW-99200-02
No leachable fillers or plasticizers
  • No glass parts to break for added safety and protection
  • Easily identify layers through translucent material
  • Easy to clean
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPTFE Drum And Tank Sampler, 7' Long
  • Warranty6 month
6 Month Warranty
Key Features
  • No glass parts to break for added safety and protection
  • Easily identify layers through translucent material
  • Easy to clean
  • 6 month warranty
More About this Item
Sample free-flowing liquids from almost any source. Safety pour top permits safe and neat transfer of contents. Polypropylene samplers have heat welded bottom caps. Push/seal bottom snaps closed easily; opens by hand or with the safety opening tool sold separately.