NOSHOK 404-FFAC NeedleValve,HardSeat Steel Female-Female 1/2" NPT Female-Female Angle
Representative image only.
NOSHOK Mfr # 404-FFAC – Item # EW-68430-19
Hard-seat design
  • Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • 10,000 psi pressure rating at 200°F (93°C)
  • Blowout-proof stem
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2\" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2\" NPT(F)
  • Orifice (in)0.438
  • Orifice (mm)11.1252
  • Cv0.44
  • Max Pressure (PSI)10,000
  • Wetted MaterialsStainless Steel
  • DescriptionNeedleValve,HardSeat Steel Female-Female 1/2\" NPT Female-Female Angle
  • Warranty3 year
3 Year Warranty
Key Features
  • Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • 10,000 psi pressure rating at 200°F (93°C)
  • Blowout-proof stem
  • 3 year warranty
More About this Item
Series 400 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.