NOSHOK 404-MFS Needle Valve, Hard Seat ss Male-Female 1/2" NPT Male-Female
Representative image only.
NOSHOK Mfr # 404-MFS – Item # EW-06292-16
Hard-seat design
  • Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • 10,000 psi pressure rating at 200°F (93°C)
  • Blowout-proof stem
PRODUCT OPTIONS
$188.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(M) x NPT(F)
  • Inlet Port1/2" NPT(M)
  • Orifice (in)0.187
  • Orifice (mm)4.7498
  • Cv0.44
  • Max Pressure (PSI)10,000
  • Wetted MaterialsStainless Steel
  • DescriptionNeedle Valve, Hard Seat ss Male-Female 1/2" NPT Male-Female
Key Features
  • Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • 10,000 psi pressure rating at 200°F (93°C)
  • Blowout-proof stem
More About this Item
Series 400 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.