NOSHOK 503-FFC Needle Valve, Soft Seat Steel Male-Female 3/8" NPT
Representative image only.
NOSHOK Mfr # 503-FFC – Item # EW-06292-39
Soft-seat design
  • Soft seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • Designed with Delrin® acetal seat
  • Straight-through porting for bidirectional, high capacity flow and easy roddable cleaning
PRODUCT OPTIONS
$66.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port3/8" NPT(F)
  • Connection TypeNPT(M) x NPT(F)
  • Inlet Port3/8" NPT(M)
  • Orifice (in)0.187
  • Orifice (mm)4.7498
  • Cv0.76
  • Max Pressure (PSI)6000@93°C
  • Wetted MaterialsStainless Steel
  • DescriptionNeedle Valve, Soft Seat Steel Male-Female 3/8" NPT
Key Features
  • Soft seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • Designed with Delrin® acetal seat
  • Straight-through porting for bidirectional, high capacity flow and easy roddable cleaning
More About this Item
Series 500 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.