NOSHOK 800-1-2-300-2 Indicating Pressure Switch, 0 to 300 psig, 0-10VDC
Representative image only.
NOSHOK Mfr # 800-1-2-300-2 – Item # EW-68455-88
  • 3 year warranty
$695.00USD / Each
$152.00 - $225.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection1/4\" NPT(M)
  • Electrical Connections4 Pin M12 x 1 Connector
  • DisplayLCD
  • Output0 to 10 VDC
  • Relays(2) NO or NC
  • DescriptionIndicating Pressure Switch, 0 to 300 psig, 0-10VDC
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
The NoShok 800 Series Electronic Pressure Switch with display provides continuous pressure monitoring and allows the programming of the set points without pressurizing. The set points, contact functions (normally open / normally closed), reset points, contact types (npn / pnp) and switching function (hysteresis / gate) are simple to adjust via the two buttons. Switching current ranging from a few u Amps to 500mA can be switched by the output transistors. By the use of time tested ceramic or thin film sensors, this pressure switch features a high level of repeatability and durability, even in the case of a high number of pressure cycles. The turnable display allows the usage of this pressure switch even under extreme installation conditions.
The high quality stainless steel housing qualifies the NoShok 800 Series also for the usage under adverse conditions. For the higher pressure ranges all wetted parts are made of stainless steel, therefore nearly every media can be used. The NoShok 800 Series is multifunctional applicable for measurement tasks within hydraulic and pneumatic applications.