NPS Spilfyter Grab & Go Base Bucket Spill Kit, Dry Formula
Representative image only.
NPS Mfr # 270006 – Item # EW-86424-51
Kit contents are packed in a 5-gallon bucket for easy portability and disposal of materials
  • Use chemical classifier to detect the presence of other chemicals
SPECS & DESCRIPTION
Specifications & Description
  • Absorption4 gal. (16 L)
  • DescriptionBase Bucket Spill Kit, Dry Formula
Includes
Three 3" x 4' hazmat socks, ten 12" x 12" hazmat pads, 10 lbs dry base neutralizer, two chemical classifier strips, one chemical classifier chart, scoop with scraper, pair of nitrile gloves, pair of goggles, disposal bag with twist tie, 5-gallon plastic bucket with snap-on lid.
Key Features
  • Use chemical classifier to detect the presence of other chemicals
More About this Item
Grab & Go buckets contain everything you need to quickly clean-up and dispose of a spill. Kit includes materials to neutralize bases.