NPS Spilfyter Grab & Go Hazmat Spill Kit, Absorbs 13 gallons, 1/Bag
Representative image only.
NPS Mfr # 211002 – Item # EW-86424-11
Kit includes everything you need for quick spill clean up, in a handy zipper bag
$273.75USD / Bag of 1
SPECS & DESCRIPTION
Specifications & Description
  • Absorption13 gal. (50 L)
  • DescriptionHazmat Spill Kit, Absorbs 13 gallons, 1/Bag
Includes
Kit includes three 3" x 10' socks, three 18" x 18" pillows, fifteen 16" x 18" pads, one pair of nitrile gloves, one pair of goggles, one disposal bag with twist tie, and one clear zipper bag.
More About this Item
Hazmat sorbents are specifically designed to absorb aggressive fluids such as acids and bases. Ideal for laboratory use.