NPS Spilfyter Wastewater Test Strips, case of 50
Representative image only.
NPS Mfr # 580050 – Item # EW-86423-52
Classify unknown spills in 30 seconds
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWastewater Test Strips, case of 50
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
Replacement test strips for Wastewater Classifier Kit.