NSI Lab Solutions FMPT-005D Qualitative Pathogens with Dairy Matrix Proficiency Test Set; 3/PK

Representative image only.
NSI Lab Solutions Item # EW-61205-03
Ensure state and accreditation regulation compliance with proficiency testing
  • Proficiency tests for qualitative identification of target microorganisms
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • For qualitative identification of S. enterica, L. monocytogenes, and E. coli O157:H7
  • Dairy matrix
  • Set of three

PRODUCT OPTIONS

$229.00USD / Pkg of 3
Order PT Express™ when you need your PT results fast

SPECS & DESCRIPTION

Specifications & Description

  • OrganismS. enterica, L. monocytogenes, and E. coli O157:H7
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid and matrix: store at room temperature
  • DescriptionQualitative Pathogens with Dairy Matrix Proficiency Test Set; 3/PK

Includes

Hydration fluid and dairy matrix.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • For qualitative identification of S. enterica, L. monocytogenes, and E. coli O157:H7
  • Dairy matrix
  • Set of three

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. Receive your final reports within 21 days of the study close date.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

A unique three-sample set contains S. enterica, L. monocytogenes, and E. coli O157:H7. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. Report present or absent for each organism of interest. Three different series available per study.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat