NSI Lab Solutions FMPT-007DB Qualitative Listeria (Nonpathogenic) with Dairy Matrix PT Express™ Set; 3/PK

Representative image only.
NSI Lab Solutions Item # UX-61205-02
On-demand proficiency test (PT) for when you need results fast
  • Proficiency tests for qualitative identification of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For qualitative identification of Listeria spp.
  • Dairy matrix
  • Set of three

PRODUCT OPTIONS

$212.25USD / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • OrganismListeria
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology PT Express
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CMatrix: store at room temperature
  • DescriptionQualitative Listeria (Nonpathogenic) with Dairy Matrix PT Express™ Set; 3/PK

Includes

Hydration fluid and dairy matrix.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For qualitative identification of Listeria spp.
  • Dairy matrix
  • Set of three

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results. Your PT report is available within 48 hours once you submit your results online.

Food Microbiology PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

This three-sample set contains nonpathogenic Listeria strain plus nonpathogenic, non-listeria organisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. Report present or absent for Listeria spp. for each sample in the set. Three different series available per study.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat