NSI Lab Solutions PH2262 Malic Acid Pharmaceutical Reference Material; 1 g

Representative image only.
NSI Lab Solutions Item # EW-05001-21
Highly purified compounds—excellent for establishing qualitative and quantitative data
  • Direct traceability of purity and identity to pharmacopeia primary standards established by multiple methods of analysis
  • Great substitute for primary standards for routine quality control purposes
  • ISO 17034 accredited

PRODUCT OPTIONS

$60.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Element/IonMalic Acid
  • FormPowder
  • Weight (g)1
  • Storage SymbolStore between 15 and 30 °C
  • CAS Number6915-15-7
  • Shelf Life (months)48
  • DescriptionMalic Acid Pharmaceutical Reference Material; 1 g

Key Features

  • Direct traceability of purity and identity to pharmacopeia primary standards established by multiple methods of analysis
  • Great substitute for primary standards for routine quality control purposes
  • ISO 17034 accredited

More About this Item

Pharmaceutical secondary reference materials have been identity and purity certified against multiple analytical methods, including HPLC, FTIR, GC, GC/MS, melting point, and titration. See the Certificate of Analysis for details of the certification process used.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat