NSI Lab Solutions SPEO-015-HERB TCLP Herbicides in Soil Proficiency Test

Representative image only.
NSI Lab Solutions Item # EW-61203-08
Ensure state and accreditation regulation compliance with proficiency testing
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • One 21 mL of spiking solution and 100 g of blank soil

PRODUCT OPTIONS

$181.00USD / Each
Order PT Express™ when you need your PT results fast

SPECS & DESCRIPTION

Specifications & Description

  • Element/Ion2,4-D, Silvex (2,4,5-TP)
  • FormLiquid
  • Volume (mL)21
  • Storage SymbolRefrigerate between 2 and 8 °C
  • UN Number1090
  • Shelf Life (months)24
  • DescriptionTCLP Herbicides in Soil Proficiency Test

Key Features

  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • One 21 mL of spiking solution and 100 g of blank soil

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Each sample contains a subset of each analyte class at concentrations exceeding regulatory levels:
2,4-D: 0 to 1.00 mg/L
Silvex (2,4,5-TP): 0 to 1.00 mg/L

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat