Nutrient broth, prepared; 1 liter bottle, 10/pk
Representative image only.
Item # EW-14204-74
Save time and money with ready-made bottles
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • A general-purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Nutrient
  • Capacity (mL)1000
  • Capacity (L)1
  • DescriptionNutrient broth, prepared; 1 liter bottle, 10/pk
  • Warranty6 month
6 Month Warranty
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • A general-purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
  • 6 month warranty
More About this Item
Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.