nVent Hoffman A606CHNFSS Stainless Steel Junction Box, Continuous Hinge, type 4X
Representative image only.
nVent Hoffman Mfr # A606CHNFSS – Item # EW-10502-72
Protects components from indoor or outdoor corrosive environments
  • Seams continuously welded and ground smooth
  • Seamless foam-in-place gasket
  • Features stainless steel screws and clamps
  • Bonding provision on door and body
  • Ratings: NEMA Type 4X, IP66
PRODUCT OPTIONS
$458.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Outside Width (in)6
  • Outside Height (in)6
  • Outside Depth (in)4
  • DescriptionStainless Steel Junction Box, Continuous Hinge, type 4X
Includes
External wall-mounting brackets. Weld nuts are provided for mounting optional panels and terminal block kits (call for details).
Key Features
  • Seams continuously welded and ground smooth
  • Seamless foam-in-place gasket
  • Features stainless steel screws and clamps
  • Bonding provision on door and body
  • Ratings: NEMA Type 4X, IP66
More About this Item
Used in either indoor or outdoor applications, these enclosures combine a rugged continuous hinge, seamless foam-in-place gasket and stainless steel screw-down clamps for a reliable seal that protects components from corrosive environments. The cover and sides of the body have a smooth #4 brushed finish.