Oakton 1618-16 Buffer, Reference Standard, pH 12.45 +/- 0.01 at 25°C (500 mL)

Representative image only.
Oakton Item # EW-86970-66
High quality, competitive pricing, and rapid service
  • NIST Traceable
  • BUFFER, pH 12.45

PRODUCT OPTIONS

$44.00USD / Each
Buffer Solutions…

SPECS & DESCRIPTION

Specifications & Description

  • Concentration12.45
  • Purity GradeComponents all ACS
  • ClassBuffers
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • Packaging500 mL PET G
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7447-40-7, 1310-73-2 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionBuffer, Reference Standard, pH 12.45 +/- 0.01 at 25°C (500 mL)

Key Features

  • NIST Traceable
  • BUFFER, pH 12.45

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat