Oakton Accumet Benchtop Ion 510 Meter with All-in-One pH/temperature electrode, 110/220 VAC

Representative image only.
Oakton Item # EW-35619-23
  • Advanced features at an economical price!
  • Store and recall 50 readings in memory
  • Feature a simple push-button keypad—ideal for use in the lab or plant.

SPECS & DESCRIPTION

Specifications & Description

  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV for ±199.9 mV; 1 mV beyond ±199.9 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Temperature compensationAutomatic or manual
  • Power (VAC)110 / 220
  • Power (Hz)50 or 60
  • Width (cm)17.78
  • Height (cm)6.0325
  • Width (in)7
  • Height (in)2 3/8
  • DescriptionAccumet Benchtop Ion 510 Meter with All-in-One pH/temperature electrode, 110/220 VAC
Conformite Europeenne

Key Features

  • Advanced features at an economical price!
  • Store and recall 50 readings in memory
  • Feature a simple push-button keypad—ideal for use in the lab or plant.

More About this Item

These easy-to-use, microprocessor-based meters feature auto-off, hold, ATC, recorder output, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. The large, dual-display LCD provides excellent visibility—even from a distance. Meters accept electrodes with a BNC connector.
Measures pH, temperature, and relative mV
Three-point pH calibration with automatic buffer recognition for both USA (pH 4.01, 7.00, 10.01) and NIST (pH 4.01, 6.86, 9.18 ) pH buffers
Automatic temperature compensation maintains reading accuracy even in fluctuating temperatures.
Recorder output—create hard copies of your data
Measures ion concentration with direct readout in ppm
Displays ppm and mV at the same time—lets you verify electrode operation while taking measurements
Calibrates at up to three ion concentration standards (choose from 0.1, 1.0, 10.0, or 100.0 ppm)
Works with all Ion Selective Electrodes with BNCconnectors

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat