This item is now discontinued. View the direct replacement: EW-35613-25.

Oakton Acorn meter with electrode, and ATC probe

Representative image only.
Oakton Item # EW-35613-00
  • High ±0.01 pH meter accuracy at economical prices
  • Convenient, compact design
  • Up to three-point push-button calibration with auto-buffer calibration
  • Displays pH and temperature with a button press

SPECS & DESCRIPTION

Specifications & Description

  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • CalibrationUp to 3 points
  • Temperature compensationAutomatic
  • Width (cm)6.985
  • Height (cm)3.4925
  • Width (in)2 3/4
  • Height (in)1 3/8
  • DescriptionAcorn meter with electrode, and ATC probe
Conformite Europeenne

Key Features

  • High ±0.01 pH meter accuracy at economical prices
  • Convenient, compact design
  • Up to three-point push-button calibration with auto-buffer calibration
  • Displays pH and temperature with a button press

More About this Item

These ph meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 100°C. The custom LCD displays temperature or pH. The pH 6 also reads mV. Meters include a protective rubber boot that also acts as meter stand for benchtop use.
All meters include: Four AAA batteries, protective rubber boot and instruction manual.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat