Oakton Acorn meter with protective rubber boot/stand Sku3561331
Representative image only.
Oakton Item # EW-35613-31
  • Direct readout of ion concentration units
  • mV measurement mode allows you to check electrode performance.
  • Compact size fits right in pocket.
  • ±500 mV range is compatible with almost all ISE electrodes.
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • CalibrationUp to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV for ±199.9 mV; 1 mV beyond ±199.9 mV
  • mV Accuracy±1 mV
  • Min ISE0.01 to 1999 ppm
  • ISE Accuracy1% of reading
  • Temperature compensationAutomatic
  • WaterproofYes
  • DescriptionAcorn meter with protective rubber boot/stand
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Direct readout of ion concentration units
  • mV measurement mode allows you to check electrode performance.
  • Compact size fits right in pocket.
  • ±500 mV range is compatible with almost all ISE electrodes.
  • 3 year warranty
More About this Item
These rugged, economical meters are very easy to use. Choose from two models. Both feature hold and auto-off functions, splash-resistant keypad and large LCD.
Meters include four AAA batteries, instructions, and a protective rubber boot which offers sturdy protection for meter and acts as meter stand for benchtop use.
Ion 6 Meter measures ion concentration (in ppm), pH, mV and °C.