Oakton All-in-One pH/ATC Electrode, Refillable, Glass, Double-Junction, BNC/Mini-Phono
Representative image only.
Oakton Item # EW-35811-74
Update, upgrade, or replace electrodes to get the best out of Oakton® meters
  • Choose from pH or ORP electrodes
  • Use pH electrode for high pH solutions and/or solutions containing high levels of sodium ions.
  • Includes 3-ft cable with BNC connector
  • ATC connector for use with Oakton pH5/6, 11/110, 510, 700, 1100/2100, or 2700 meters
  • 1 year warranty
PRODUCT OPTIONS
$265.50USD / Each
$295.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRefillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionAll-in-One pH/ATC Probe,Refillable/DJ/Glass; BNC/Mini-Phono
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from pH or ORP electrodes
  • Use pH electrode for high pH solutions and/or solutions containing high levels of sodium ions.
  • Includes 3-ft cable with BNC connector
  • ATC connector for use with Oakton pH5/6, 11/110, 510, 700, 1100/2100, or 2700 meters
  • 1 year warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.