Oakton by Cole-Parmer® In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Plug/BNC
Representative image only.
Cole-Parmer Item # EW-27001-70
Integral, one-piece electrode and housing for installation convenience
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
  • 1 year warranty
PRODUCT OPTIONS
$245.00USD / Each
BUY MORE & SAVE
  • 1-5$245.00USD / Each
  • 6-11$220.50USD / Each
  • 12-23$208.25USD / Each
  • 24+$196.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)13
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • DescriptionIn-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Plug/BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
  • 1 year warranty
More About this Item
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.
Note: 100 Ω RTD ATC elements are platinum and conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ