Oakton by Cole-Parmer® pH electrode, in-line, single-junction, no ATC, 10-ft cable, BNC
Representative image only.
Cole-Parmer Item # EW-05993-90
Integral, one-piece electrode and housing for installation convenience
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
  • 1 year warranty
PRODUCT OPTIONS
$165.00USD / Each
BUY MORE & SAVE
  • 1-5$165.00USD / Each
  • 6-11$148.50USD / Each
  • 12-23$140.25USD / Each
  • 24+$132.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionpH electrode, in-line, single-junction, no ATC, 10-ft cable, BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
  • 1 year warranty
More About this Item
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.
Note: 100 Ω RTD ATC elements are platinum and conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ