Oakton Conductivity and TDS Standard, 2070 µS; 500 mL

Representative image only.
Oakton Item # EW-00653-27
Ideal for laboratory and field analysis
  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • Use when readings are expected to be around 2070 µS/cm

PRODUCT OPTIONS

SPECS & DESCRIPTION

Specifications & Description

  • Conductivity Value (µS)2070
  • Conductivity Value (mS)2.07
  • Volume (mL)500
  • Volume (oz)16.9
  • Conductivity-to-TDS Calibration Values (ppm 442)1500
  • Conductivity-to-TDS Calibration Values (ppm KCl)1045
  • Conductivity-to-TDS Calibration Values (ppm NaCl)1041
  • DescriptionConductivity and TDS Standard, 2070 µS; 500 mL

Key Features

  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • Use when readings are expected to be around 2070 µS/cm

More About this Item

KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60-mL bottle, but included in insert). Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60-mL bottle, but included in insert). Manufactured under ISO 9000 quality guidelines and traceable to NIST standards. If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months. Conductivity options range from 1 µS/cm to 200,000 µS/cm.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat