Environmental Express DO300 Replacement DO Sensor Cap
Representative image only.
Environmental Express Item # EW-35661-00
Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • Replacement DO sensor cap for 35660-96 and -98 DO sensors
  • 6 month warranty
PRODUCT OPTIONS
$182.00USD / Each
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeReplacement DO Sensor Cap
  • Range0 to 20 mg/L
  • MaterialPVC and PMMA
  • DescriptionReplacement DO Sensor Cap
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • Replacement DO sensor cap for 35660-96 and -98 DO sensors
  • 6 month warranty
More About this Item
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.