Oakton meter with single-junction pH/ATC probe Sku3561813

Representative image only.
Oakton Item # EW-35618-13
Waterproof and dustproof with IP67-rated housing that floats!
  • Built-in memory function makes it easy to recall data
  • Five-point pH calibration with auto-buffer recognition
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)8.89
  • Height (cm)18.7325
  • Width (in)3 1/2
  • Length (in)1 3/4
  • Height (in)7 3/8
  • Descriptionmeter with single-junction pH/ATC probe
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Key Features

  • Built-in memory function makes it easy to recall data
  • Five-point pH calibration with auto-buffer recognition
  • 3 year warranty

More About this Item

Advanced pH 310 Meter has a real-time clock that lets you stamp 50 stored readings and calibration data with date/time — ideal for Good Laboratory Practice (GLP) standards. The pH 310 also features selectable automatic endpoint (freezes reading when stable), and selectable temperature (°C or °F).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat