This item is now discontinued. View the direct replacement: EW-35421-02.

Oakton meter without probes Sku3562102

Representative image only.
Oakton Item # EW-35621-02
  • Selectable 0.001/0.01 pH resolution with ±0.002/0.01 pH accuracy
  • Two-line backlit display provides clear operating instructions
  • Store and recall up to 100 sets of data—send data to a computer via the RS-232 port or to a recorder via the mV output

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV for ±199.9 mV; 1 mV beyond ±199.9 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0.001 to 19999 ppm
  • ISE Accuracy±0.5% of reading
  • Temperature compensationAutomatic or manual
  • Power (VAC)110 / 220
  • Power (Hz)50 or 60
  • Width (cm)17.78
  • Height (cm)6.0325
  • Width (in)7
  • Height (in)2 3/8
  • Descriptionmeter without probes

Key Features

  • Selectable 0.001/0.01 pH resolution with ±0.002/0.01 pH accuracy
  • Two-line backlit display provides clear operating instructions
  • Store and recall up to 100 sets of data—send data to a computer via the RS-232 port or to a recorder via the mV output

More About this Item

Meters feature a two-line, backlit display that shows the last calibration date/time and provides clear instructions or setup and calibration. Meet GLP requirements—meters mark all calibration and stored data with time/date stamps. Nonvolatile memory stores up to 100 data sets to simplify documenting large volumes of critical ISE (2100 models only), pH, or mV measurements.

Meters automatically calculate and display electrode slope and offset—ideal for checking electrode quality. Manually send stable readings to your printer or PC at the touch of a button or program the meter to send automatically at user-defined intervals. Organize data files for analysis with the included Windows® based DAS software. Quick-reference card attached to underside of meter keeps commonly used procedures readily at hand. Meters accept electrodes with BNC connections.

pH 1100 Meter Features
Meters feature selectable manual or automatic temperature compensation.
Calibrate using select NIST/US standard or custom buffer sets
User-defined calibration alarm sign

pH/Ion 2100 Meter Features
All features of the pH 1100 meter plus:
Ion concentration mode directly displays readings in ppm, ppt, percent, or any other units of your choice.
w Customize up to five calibration points

What’s included: meter, electrode holder, software, quick-reference card, and 110/220 VAC adapters. pH/Ion 2100 models also include a pH electrode and an ATC probe.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat