Oakton Optical Dissolved Oxygen (RDO) Calibration Chamber/Sleeve
Representative image only.
Oakton Item # UX-35640-58
Quality accessories for Oakton RDO Meters
  • 1 year warranty
PRODUCT OPTIONS
$231.00USD / Each
Include Accessories Today!
Oakton® DO 450 and pH/DO 450 Meter (RDO) Accessories
ITEM(S) ON SALE
$231.00 - $1,784.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Calibration
  • DescriptionOptical Dissolved Oxygen (RDO) Calibration Chamber/Sleeve
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Oakton calibration chamber is threaded to fit the RDO 450 probe, and is good for keeping the probe hydrated during storage.