Oakton ORPTestr Replacement ORP Electrode Sensor
Representative image only.
Oakton Item # EW-35650-09
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample
  • Choose from a variety of accessories for Oakton® meters
  • 6 month warranty
PRODUCT OPTIONS
$157.50USD / Each
$175.00USD / Each
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$39.00 - $405.00USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton ORPTestr 10, 35650-10
  • DescriptionORPTestr Replacement ORP Electrode Sensor
  • Warranty6 month
6 Month Warranty
Key Features
  • Choose from a variety of accessories for Oakton® meters
  • 6 month warranty
More About this Item
For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.