Oakton ORPTestr® Waterproof Pocket ORP Tester
Representative image only.
Oakton Item # EW-35650-14
Use with any standard ORP electrode with BNC connector
  • Ideal for checking chromate reduction, cyanide oxidation, and other ORP readings
  • Large, horizontal readout
  • IP67-rated housing floats in water and sits flat on a hard surface
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeTester
  • Display typeLCD
  • ConnectionsBNC and WP Mini-din
  • Min mV (mV)-1000
  • Max mV (mV)1000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±2 mV
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • DescriptionWaterproof Pocket ORP Tester
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier
Key Features
  • Ideal for checking chromate reduction, cyanide oxidation, and other ORP readings
  • Large, horizontal readout
  • IP67-rated housing floats in water and sits flat on a hard surface
  • 1 year warranty
More About this Item
ORPTestr® 10 BNC allows you the convenience of using any ORP electrode with a BNC connection. Order electrodes separately.
Use this versatile instrument for applications ranging from swimming pool testing to pulp bleaching. Meter features ±2 mV accuracy and a ±150 mV offset. Housing allows you to attach a lanyard (included) for easy retrieval. Additional features include a hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.