This item is now discontinued. We recommend the following as suggested replacements: EW-35419-55

Oakton pH 2700 Benchtop Meter with Electrode Arm and NIST-Traceable Calibration

Representative image only.
Oakton Item # EW-35420-23
Affordable meters that meet GLP requirements!
  • Oversize, informative display with bright backlight
  • High 0.001 pH resolution with ±0.002 pH accuracy
  • Store and recall up to 500 sets of data—send data to a PC or printer
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • InterfaceRS-232
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Height (in)2 3/4
  • DescriptionBenchtop Meter with electrode arm and NIST-traceable Calibration
  • Warranty3 year
3 Year Warranty

Key Features

  • Oversize, informative display with bright backlight
  • High 0.001 pH resolution with ±0.002 pH accuracy
  • Store and recall up to 500 sets of data—send data to a PC or printer
  • 3 year warranty

More About this Item

More features of the Oakton 2700 metersOakton 2700 meters meet GLP requirements—meters stamp all calibration and stored data with time/date. Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH or mV measurements. Meters automatically calculate and display electrode slope and offset—ideal for checking electrode quality. Order RS-232 and optional RS-232 to USB adapter separately.

Advanced set-up options allow for a large degree of user customization. Datalogging intervals can be programmed into the meter for convenient continuous monitoring. Programmable limit alarm alerts you when readings fall out of range. Calibrate up to 6 points for pH using USA, NIST, DIN, or custom buffer sets. Optional password protection prevents changes to meter setup and calibration.

Oversized 3-1/4" x 2-7/16" (8.3 x 6.2 cm) graphic LCD can be viewed from across the room. Bright backlight and large digits allow readings in all conditions. Informative display shows pH or Redox measurements together with temperature, electrode status, cal points, time and date all at once! Cal-due alarm lets you know when recalibration is necessary for measurement accuracy. Visual stability indicator informs you when measurements have stabilized, eliminating guesswork. Comprehensive self-diagnostics make troubleshooting a breeze.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat