Cole-Parmer Epoxy-Body pH Electrode with ATC, Large Ceramic Junction, Gel-Filled; 1/EA
Representative image only.
Cole-Parmer Item # EW-05992-14
Combine durability with reliability
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • Compatible with 35613-XX Series Acorn 5 pH meter, and others
  • 30K thermistor ATC with 2.5 mm audio (phono) plug for temperature compensation
PRODUCT OPTIONS
$180.90USD / Each
$201.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationspH
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionCeramic
  • Reference Electrolyte4M KCl, polymerized
  • Min pH (pH)0
  • Max pH (pH)14
  • ATC Connector2.5 mm audio
  • ATC Element30K thermistor
  • ConnectorBNC
  • DescriptionEpoxy-Body pH Electrode with ATC, Large Ceramic Junction, Gel-Filled; 1/EA
Includes
3.3-ft (1-m) cable with BNC connector.
Key Features
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • Compatible with 35613-XX Series Acorn 5 pH meter, and others
  • 30K thermistor ATC with 2.5 mm audio (phono) plug for temperature compensation
More About this Item
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Oakton Basic conductivity meter
4.0
(based on 1 review)
Reviewed by 1 customer
Displaying review 1
Back to top
4.0
Convenient "All-in-one"
By 
from Michigan
About Me Lab Assistant
Verified Reviewer
Pros
  • Accurate
  • Easy To Use
  • Works Properly
Cons
    Best Uses
    • In The Field
    • In The Lab
    • Research
    Comments about Oakton Basic conductivity meter:
    Very convenient to have both the temperature and pH in the same probe.
    Displaying review 1
    Back to top
    Q&A
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      does it read mv?
      Asked on 9/20/2010 by Anonymous
      1 answer
      • STAFF EXPERT
        A:
        The electrode does give readings in a mv form. however, the reading is based off the meter that you use. As long as your meter reads in mv mode then you will be able to read in that form with this electrode.
        Answered on 9/22/2010 by Mike from Tech Support
    Displaying question 1
    Do you have a question about this product? 