Oakton pH Electrode, Single-Junction, Gel, BNC Connector
Representative image only.
Oakton Item # EW-59001-65
Combine durability with reliability
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • 6 month warranty
PRODUCT OPTIONS
$99.50USD / Each
BUY MORE & SAVE
  • 1-3$99.50USD / Each
  • 4+$89.55USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationspH
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionpH electrode, single-junction, gel, BNC connector
  • Warranty6 month warranty
6 Month Warranty
Includes
3.3-ft (1-m) cable with BNC connector.
Key Features
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
  • 6 month warranty
More About this Item
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.