Oakton pH Spear Waterproof Pocket pH Testr™

Representative image only.
Oakton Item # EW-35634-40
Versatile, convenient, and accurate spear-tip pH tester
  • Electrode junction is 200 times larger than standard electrodes, making it virtually impossible to clog
  • Rugged spear tip makes it ideal for soft solids and semisolid materials like meats, cheeses, and fruit
  • Long, thin electrode can be used to determine pH in test tubes and other narrow containers
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationManual
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • DescriptionWaterproof Pocket pH Testr™
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Electrode junction is 200 times larger than standard electrodes, making it virtually impossible to clog
  • Rugged spear tip makes it ideal for soft solids and semisolid materials like meats, cheeses, and fruit
  • Long, thin electrode can be used to determine pH in test tubes and other narrow containers
  • 1 year warranty

More About this Item

The unique junction design provides increased electrolyte flow, resulting in greater contact between sample and reference, ensuring fast, stable readings. The double-junction design offers enhanced performance. Additional features include a hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat