Environmental Express PH300 pH Sensor Head; 5-m Cable
Representative image only.
Environmental Express Item # EW-35661-01
Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
PRODUCT OPTIONS
$319.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypepH Sensor Head
  • Range—2 to 20 pH
  • MaterialABS plastic and polycarbonate
  • Length (cm)8.5
  • DescriptionpH Sensor Head; 5-m Cable
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Suitable for use with Oakton 300 series smart meters and meter kits
  • 6 month warranty
More About this Item
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.