Oakton pHCON Calibration Kit with Carrying Case
Representative image only.
Thermo Scientific Item # EW-35630-69
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample
  • Choose from a variety of accessories for Oakton® meters
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton pH/Con 300
  • DescriptionpHCON Calibration Kit with Carrying Case
  • Warranty6 month
6 Month Warranty
Includes
calibration solution pouches (three each of pH 4.01, 7.00, 10.00 and two each of conductivity 447 μS, 1413 μS, 2764 μS, 15,000 μS); three rinse water pouches; sample bottle; and hard carrying case
Key Features
  • Choose from a variety of accessories for Oakton® meters
  • 6 month warranty
More About this Item
For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.