Oakton pHTestr® 30 Waterproof Pocket Tester with Calibration Kit

Representative image only.
Oakton Item # EW-35634-90
Versatile, compact design works in a field or laboratory setting
  • 0.01 pH accuracy along with dual pH and selectable C or F temperature display
  • A complete meter with key features at an economical price
  • Replaceable, double-junction electrode sensors save time and money
  • IP67-rated housing sits flat on the table and floats in water
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Diameter (cm)3.81
  • Diameter (in)1 1/2
  • DescriptionWaterproof Pocket Tester with Calibration Kit
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • 0.01 pH accuracy along with dual pH and selectable C or F temperature display
  • A complete meter with key features at an economical price
  • Replaceable, double-junction electrode sensors save time and money
  • IP67-rated housing sits flat on the table and floats in water
  • 1 year warranty

More About this Item

Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox sensor housing resists most chemicals.

The pHTestr 30 has a replaceable sensor for easy maintenance. pHTestr 30 purchased before Oct. 2019 use replacement sensor 35624-38. pHTestr 30 purchased after Oct. 2019 use replacemenr sensor 35634-03. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage.

The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat