Oakton Reagent Kit for Colorimeters, Chlorine Dioxide Testing; 100 Tests/Kit
Representative image only.
Oakton Item # EW-35645-68
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Chlorine Dioxide (Glycine) Reagent Kit — enough for 100 tests
  • Use with the C-103 Oakton Colorimeter
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersChlorine dioxide
  • Number of Tests Included100
  • DescriptionReagent Kit for Colorimeters, Chlorine Dioxide Testing; 100 Tests/Kit
Key Features
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Chlorine Dioxide (Glycine) Reagent Kit — enough for 100 tests
  • Use with the C-103 Oakton Colorimeter