Oakton Reference fill solution, 4 M KCl, 125 mL bottle
Representative image only.
Oakton Item # EW-35803-74
Optimize water analysis
  • Choose from a variety of accessories
  • 1 year warranty
PRODUCT OPTIONS
$39.00USD / Each
Include Accessories Today!
Oakton® Water Analysis Accessories
ITEM(S) ON SALE
$38.00 - $222.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton double junction refillable pH electrodes
  • DescriptionReference fill solution, 4 M KCl, 125 mL bottle
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.