Oakton Rugged pH Electrode,12x110mm, Glass Body, Double Junction, Refill
Representative image only.
Oakton Item # EW-55510-38
Affordable, high-accuracy pH electrodes for every application
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
PRODUCT OPTIONS
$225.00USD / Each
$250.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRugged, refillable
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)110
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)110
  • Cable Length (m)1
  • DescriptionRugged pH Electrode,12x110mm, Glass Body, Double Junction, Refill
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Cole-Parmer high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility over entire pH range. Fill solution purchased seperately, 05992-47 for single junction, 05992-48 for double juncion electrodes