Oakton Waterproof pH Electrode with ATC, Single-Junction, Polycarbonate-Body, Gel-Filled
Representative image only.
Oakton Item # EW-35808-71
Oakton offers a variety of pH electrodes for your general purpose and speciality laboratory applications
  • Choose single-junction electrodes to measure pH in clean aqueous samples
  • Choose double-junction electrodes for use with samples that may be contaminating with particulates, heavy metals or organic compounds
  • Replacement All in One electrode for meter series 150 & 450
  • 1 year warranty
PRODUCT OPTIONS
$132.30USD / Each
$147.00USD / Each
Often Bought with…
Ohaus MB90 Moisture Determination Balance, 90 g X 0.001 g
$4,382.56USD / Each
$5,096.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGel-filled
  • Temperature CompensationYes
  • ATC Connector6-pin waterproof
  • ConnectorBNC
  • DescriptionWaterproof pH Electrode with ATC, Single-Junction, Polycarbonate-Body, Gel-Filled
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose single-junction electrodes to measure pH in clean aqueous samples
  • Choose double-junction electrodes for use with samples that may be contaminating with particulates, heavy metals or organic compounds
  • Replacement All in One electrode for meter series 150 & 450
  • 1 year warranty
More About this Item
Low-maintainance epoxy-bodied electrodes provide the durability for field work. Refillable electrodes provide greater accuracy and faster response for more critical analytical work.
Compatible with Oaktoh pH 300/310, pH/CON 300, pH/CON 150, & 450 series meters.