Oakton Temp-360 Datalogging Precision RTD Thermometer with Boot
Representative image only.
Oakton Item # EW-91426-60
Log up to 2,000 sets of readings in real time with time and date stamp
  • 3-way hands-free operation
  • Transfer data via USB
  • Menu driven set up to easily customize your meter
  • Rugged armor—built to withstand the toughest conditions.
  • Probes are sold separately
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-330
  • Max Temperature (° F)2210
  • Min Temperature (° C)-201
  • Max Temperature (° C)1210
  • Accuracy±0.06°F (±0.03°C); ±0.1°F (±0.1°C) below -100°; ±1.0°F (±1.0°C) above 1000°
  • Resolution0.01°; 0.1° between -100.0° to -330° and 100.0° to 999.9°; 1° above 1000°
  • OutputUSB
  • Data LoggingYes
  • Memory Capacity2000 data points
  • IP RatingIP54
  • Low Battery IndicatorYes
  • Display Type1.5\" Display, 4-digit LCD
  • Height (in)7
  • Battery Life400 hours
  • BatteryAA Battery (3)
  • DescriptionTemp-360 Datalogging Precision RTD Thermometer with Boot
  • Warranty3 year
3 Year Warranty
Key Features
  • 3-way hands-free operation
  • Transfer data via USB
  • Menu driven set up to easily customize your meter
  • Rugged armor—built to withstand the toughest conditions.
  • Probes are sold separately
  • 3 year warranty
More About this Item
Choose the Oakton® Temp-340 thermometer that has real time datalogging and a USB data output. This robust and reliable meter is ideal for use in food processing plants, research and science labs, building maintenance areas and more! The sturdy armored construction protects against knocks and jars. Ergonomically designed for both large and small hands alike. The sealed keypad and ABS case meet IP54 standards for splash resistance. The meter has a large illuminated display, and a seven-button control panel that allows the user through easy-to-use menus to choose desired functionality.
Features include real-time datalogging and a USB data output. The sevenbutton control panel and intuitive on-screen menus guide you in choosing your desired functionality. Additional meter features include Min/Max, Hold, and average functions; °F/°C selection; time stamp/clock; countdown timer; out-of-range alarm with audible/visible indicator; auto shutoff; and low-battery indicator. Order an optional AC adapter to conserve battery life during extended operations. Manually store or automatically log up to 2000 sets of readings in real time. Log interval is selectable from 1 second to 60 minutes. (Meter requires a RTD probe with a Oakton three-pin plug)
OAKTON’s exclusive “3-WAY HANDS-FREE” capability allows “hands-free” operation— use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meter comes standard with the integral stand; the “hands-free” option adds magnets and strap (order separately)