ODE 21IN7KOV350-IH Solenoid Valve, 316 SS, with 1 1/4 NPT(F) Port
Representative image only.
ODE Mfr # 21IN7KOV350-IH – Item # EW-08610-48
ATEX-rated explosion-proof coils are designed for petrochemical and other industrial applications
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications
PRODUCT OPTIONS
$1,041.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1.25
  • Outlet Port1-1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1-1/4" NPT(F)
  • Orifice (in)1.38
  • Orifice (mm)35.052
  • Cv79.25
  • Power (VAC)120
  • Max Temperature (° F)176
  • Max Temperature (° C)80
  • Wetted MaterialsStainless Steel
  • DescriptionSolenoid Valve, 316 SS, with 1 1/4 NPT(F) Port
Key Features
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications