ODE 21IN8KOV400-IH Solenoid Valve, 316 SS, with 1 1/2 NPT(F) Port
Representative image only.
ODE Mfr # 21IN8KOV400-IH – Item # EW-08610-50
ATEX-rated explosion-proof coils are designed for petrochemical and other industrial applications
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications
PRODUCT OPTIONS
$1,082.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1.5
  • Outlet Port1-1/2" (NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1-1/2" NPT(F)
  • Orifice (in)1.57
  • Orifice (mm)39.878
  • Cv89.82
  • Power (VAC)120
  • Max Temperature (° F)176
  • Max Temperature (° C)80
  • Wetted MaterialsStainless Steel
  • DescriptionSolenoid Valve, 316 SS, with 1 1/2 NPT(F) Port
Key Features
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications