Ohaus 183-00 Rod and Clamp for Under-Balance Weighing
Representative image only.
Ohaus Mfr # 183-00 – Item # EW-11609-67
Expand the uses of your balance with the below-balance weighing hook
  • 1 year warranty
PRODUCT OPTIONS
$89.18USD / Each
$98.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRod and Clamp for Under-Balance Weighing
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A hook is available underneath all balances for weighing beneath the balance. This is particularly useful in applications such as determining the measurment of large pieces.