Ohaus CKW55 Champ Washdown Checkweighing Indicator, Ip66, NSF/Usda/NTEP Listed
Representative image only.
Ohaus Mfr # CKW55 – Item # EW-90453-18
Advanced indicators for food and industrial applications
  • RS232 included with GMP and Statistics output
  • Optional second RS232, RS485/422 outputl
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Weighing Units%, kg, g, lb, oz, B/G
  • DescriptionChamp Washdown Checkweighing Indicator, Ip66, NSF/Usda/NTEP Listed
  • Warranty1 Year
1 Year Warranty
Key Features
  • RS232 included with GMP and Statistics output
  • Optional second RS232, RS485/422 outputl
  • 1 year warranty
More About this Item
The OHAUS CKW is the advanced all stainless steel checkweighing indicator with quick set-up of targets and full configurability. Can be set-up to checkweigh by weight or as a percentage of reference weight with full control over how progress is shown -- even display progress using 24 LED yellow/green/red bar without any display of weight on main LED display! The Ohaus CKW-55 indicator provides IP66/NEMA 4X washdown protection, is NSF listed/Certified, USDA/AMS accepted, and supports HACCP-Certified Systems