Omegadyne LC501-200 SS Cantilever Beam, Cable Style; 200 lb

Representative image only.
Omegadyne Mfr # LC501-200 – Item # EW-59890-36

$504.75USD / Each

$162.00USD / Each

for an additional
$162.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (lbs)200
  • Capacity (kg)91
  • Output3 mV/V ±0.25%
  • DescriptionSS Cantilever Beam, Cable Style; 200 lb

More About this Item

Stainless steel cantilever beams

  • FM Intrinsically Safe
  • All Stainless Steel for Harsh Industrial Applications
  • 0.25% Interchangeability for Multiple Load Cell Applications
  • 5-Point Calibration Provided

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat