Omegadyne LCGD-2K Load Cell, 2000 lb capacity, Mini Compression Load Cell
Representative image only.
Omegadyne Mfr # LCGD-2K – Item # EW-59890-58
$674.25USD / Each
$154.00USD / Each
for an additional
$154.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)2000
  • Capacity (kg)909
  • Output2 mV/V nominal
  • DescriptionLoad Cell, 2000 lb capacity, Mini Compression Load Cell
More About this Item
Miniature industrial load cell These small low-profile, compression load cells have excellent long-term stability. An all stainless steel construction assures reliability in severe industrial environments. These cells are designed to operate by mounting on a flat surface, and they have a load button machined as an integral part of their basic design.