Omegadyne LCGD-5K Load Cell 5000LB Capacity Mini Compression Load Cell
Representative image only.
Omegadyne Mfr # LCGD-5K – Item # EW-93955-75
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)5000
  • Capacity (kg)2269
  • Output2 mV/V nominal
  • DescriptionLoad Cell 5000LB Capacity Mini Compression Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Miniature industrial load cell These small low-profile, compression load cells have excellent long-term stability. An all stainless steel construction assures reliability in severe industrial environments. These cells are designed to operate by mounting on a flat surface, and they have a load button machined as an integral part of their basic design.