Omegadyne LCHD-25 Load Cell25LB.Cap Low Profile Pancake Tension And Compression Load Cell
Representative image only.
Omegadyne Mfr # LCHD-25 – Item # EW-93955-82
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)25
  • Capacity (kg)11
  • Output=25 lb: 2 mV/V ±0.25%; =50 lb: 3 mV/V ±0.25%
  • DescriptionLoad Cell25LB.Cap Low Profile Pancake Tension And Compression Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Low profile pancake style
  • 0.25% interchangeability for scale applications
  • All stainless steel construction
  • Heavy-Duty design
  • High accuracy
  • Low profile for easy installation