Omni International 15001 Mixer Homogenizer System Adapter for Rotor-Stator Generators
Representative image only.
Omni International Mfr # 15001 – Item # EW-04729-58
Required for use with an Omni Rotor-Stator Generator in an open container
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Use with Generators04729-26, 04729-28, 04729-38
  • DescriptionMixer Homogenizer System Adapter for Rotor-Stator Generators
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Adapter, required to use rotor-stator generators in open containers.