Omron F3SN-A0457P25 Safety Light Curtain, 13 beam - 217 mm Protective height
Representative image only.
Omron Mfr # F3SN-A0457P25 – Item # EW-49801-04
Provide protection against category 4 hazards
  • Meets ANSI/RIA, OSHA, IEC, EN, UL, and CSA standards
  • Units are self-contained – no control box required
  • Optional setting console makes setup/operation simple
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year WarrantyUL Listed to Canadian safety standards.IP 65UL Listed.
Key Features
  • Meets ANSI/RIA, OSHA, IEC, EN, UL, and CSA standards
  • Units are self-contained – no control box required
  • Optional setting console makes setup/operation simple
  • 1 year warranty
More About this Item
Use these independently certified light curtains for protection against life-threatening category 4 hazards. These light curtains are available in incremental sizes, by number of beams, so you can specify the exact protective height you need saving on each system. Indicator LED’s show status, error, and light intensity on both the emitter and the receiver. The slim profile is space efficient and ideal for applications requiring category 4 hazard protection. Series connectable models are available upon request, allowing for up to three sets of 240 beams to be connected in series, and provide an auto and manual reset function.
Use your own, or order the optional safety relay, and cables, (below) to complete your light curtain system, and reduce wiring time and eliminate possible improper wiring. The optional setting console may be added to provide setup and operation flexibility. The fixed blanking of one to nine beams, one to three zones, and a teach function offer design flexibility that will meet your installation needs. Floating blanking of one to three beams, and the teach function provide a sequential mode ensuring that beams have to be broken consecutively (floating and fixed blanking requires optional setting console). After powering the safety light curtain system on the diagnostic function feature performs a complete self-test of the light curtain system within one second, and performs a self-test periodically during operation.
What’s included: One each emitter and receiver light curtain devices.